missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Growth Hormone Releasing Peptide-2

$654.60 - $654.60

Specifications

CAS 158861-67-7
Content And Storage Store at -20°C
Format Solid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66029MB
View Documents
Thermo Scientific™
J66029MB
25 mg
Each for $654.60
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

158861-67-7
Store at -20°C
Solid
Videos
SDS
Documents

Documents