missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Growth Hormone Releasing Peptide-2

Catalog No. AAJ66029MB
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66029MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66029MB Supplier Thermo Scientific™ Supplier No. J66029MB

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 158861-67-7
Content And Storage Store at -20°C
Format Solid
Quantity 25 mg