missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Ground Glass Joint Conical Reaction Vial

Joint: 14/20.

$41.68 - $47.11

Specifications

Color Clear
Material Borosilicate Glass
Sterility Non-sterile
Disposable Single-use
For Use With (Application) Organic Synthesis
View More Specs

Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
31502112
View Documents
Synthware™
V141003
3 mL
Each for $41.68
Only null left
 
31502113
View Documents
Synthware™
V141005
5 mL
Each for $42.39
Only null left
 
31502114
View Documents
Synthware™
V141008
8 mL
Each for $47.11
Only null left
 
Description

Description

Specifications

Specifications

Clear
Non-sterile
Organic Synthesis
Heavy-wall
No
Borosilicate Glass
Single-use
Yes
1 Ea.
Videos
SDS
Documents

Documents