UserName
Recombinant mouse FER (542-end) was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.