Learn More
SignalChem Biotech FYN C, Unactive 1000ug
Catalog No. 502574856
Encompass
Recombinant full-length human FYN C was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 502574856 Supplier SignalChem Biotech Supplier No. F1514G1000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant full-length human FYN C was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.