missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Griseofulvin 97.0+%, TCI America™
Supplier: TCI America G038425G
Specifications
| (+)-Griseofulvin | |
| 220°C | |
| C17H17ClO6 | |
| 25 g | |
| DDUHZTYCFQRHIY-RBHXEPJQSA-N | |
| (2S,5'R)-7-chloro-3',4,6-trimethoxy-5'-methylspiro[1-benzofuran-2,4'-cyclohex-2-ene]-1',3-dione | |
| 441140 | |
| 352.77 | |
| Crystalline Powder |
| 126-07-8 | |
| White | |
| MFCD00082343 | |
| griseofulvin, +-griseofulvin, amudane, fulvicin, griseofulvina, griseofulvinum, grisactin, grisefuline, grisovin, griseofulvine | |
| CC1CC(=O)C=C(C12C(=O)C3=C(O2)C(=C(C=C3OC)OC)Cl)OC | |
| 352.767 | |
| CHEBI:27779 | |
| ≥97.0% (HPLC) |
Chemical Identifiers
| 126-07-8 | |
| 352.767 | |
| DDUHZTYCFQRHIY-RBHXEPJQSA-N | |
| 441140 | |
| (2S,5'R)-7-chloro-3',4,6-trimethoxy-5'-methylspiro[1-benzofuran-2,4'-cyclohex-2-ene]-1',3-dione |
| C17H17ClO6 | |
| MFCD00082343 | |
| griseofulvin, +-griseofulvin, amudane, fulvicin, griseofulvina, griseofulvinum, grisactin, grisefuline, grisovin, griseofulvine | |
| CHEBI:27779 | |
| CC1CC(=O)C=C(C12C(=O)C3=C(O2)C(=C(C=C3OC)OC)Cl)OC |
Safety and Handling
EINECSNumber : (8)-0462&(9)-0336
RTECSNumber : WG9800000
TSCA : No
Recommended Storage : Refrigerator