missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Grips™ Premium Goatskin Leather Drivers Gloves

Catalog No. 1146264F
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
1146264F X-Large
1146264C Small
1146264D Medium
1146264E Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 1146264F Supplier Wells Lamont™ Supplier No. Y0769XL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Goatskin leather is soft, pliable, and longer lasting than cow and pig leather drivers. Grips™ Premium Goatskin Leather Drivers Gloves are ideal for use in warehousing, construction, fork lift driving, and oil and gas industries.

  • Top quality goatskin with natural added lanolin for softness and extended wear
  • Goatskin has a higher natural tensile strength despite abrasion
  • Gunn cut and keystone thumb for added dexterity and flexibility
  • Bound hem

Specifications

Color Beige
Grade Premium
Hand Specific
Material Goatskin Leather
Product Type General Purpose Gloves
Size X-Large
Style Gunn
Glove Cuff Style Open
Hem Color Blue
Lining Unlined
Series Y0769
Thumb Style Keystone
Show More Show Less