missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One MiniCollect™ Capillary Blood Collection System Accessories
Description
- Capillary or funnel can be inserted into MiniCollect tubes without removing the cap
- 13 x 75 mm carrier tubes facilitate bar code labeling, centrifugation, and storage
- Use amber carrier tube for sheilding light-sensitive analytes
Specifications
Specifications
| Color | Red |
| Product Type | Serum Tube |
| Capacity (Metric) | 80 μL |
| For Use With (Equipment) | Greiner Vacuette MiniCollect Capillary Blood Collection System |
| Tube Size | 80 μL |