missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One MiniCollect™ Capillary Blood Collection System Accessories

Catalog No. 22040083
Change view
Click to view available options
Product Type:
Capillary Tube
Funnel
Serum Tube
Tube Size:
N/A
80 μL
13 x 75 mm
Color:
Amber
Clear
N/A
Red
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Tube Size Color
22040083 Funnel N/A N/A
22040082 Capillary Tube 13 x 75 mm Clear
22040081 Capillary Tube 13 x 75 mm Amber
22040090 Serum Tube 80 μL Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22040083 Supplier Greiner Bio-One Supplier No. 450421

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with MiniCollect Capillary Blood Collection System

  • Capillary or funnel can be inserted into MiniCollect tubes without removing the cap
  • 13 x 75 mm carrier tubes facilitate bar code labeling, centrifugation, and storage
  • Use amber carrier tube for sheilding light-sensitive analytes

Specifications

Color N/A
Product Type Funnel
For Use With (Equipment) Greiner Vacuette MiniCollect Capillary Blood Collection System
Tube Size N/A