missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Luer Adapter

Catalog No. 22040000
Change view
Click to view available options
Product Type:
Luer Adapter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22040000 Luer Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22040000 Supplier Greiner Bio-One Supplier No. 450070

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with Safety/Blood Collection sets or Luer needles in routine blood collection

  • Sterile, single-use device
  • Manufactured from stainless steel and polypropylene
  • Fitted with a safety valve at one end
  • Perforated label simplifies identification and serves as a seal of integrity

Specifications

Product Type Luer Adapter
Material Stainless Steel, Polypropylene
Sterility Sterile
Certifications/Compliance CE, OSHA
Disposable Single-use
For Use With (Equipment) Safety/Blood collection sets or 20G luer needles in routine Blood collection
Includes Safety Valve at one end
Storage Requirements 4°C to 25°C