Learn More
Greiner Bio-One Plasma Tubes with Lithium Heparin and Gel
For determinations in clinical chemistry and immunochemistry. Greiner Bio-One Plasma Tubes with Lithium Heparin and Gel includes a safety pull cap.
Supplier:  Greiner Bio-One 456087P
 View more versions of this product
Catalog No. 22040540
Only null left
Description
Description
Plasma stability in a spun tube.
  • Interior tube wall is coated with lithium heparin, an anticoagulant that binds antithrombins to block the coagulation cascade producing a plasma sample
  • Contain a barrier gel in the tube that moves upward to form a stable barrier separating plasma from cells during centrifugation; plasma may be aspirated directly from the tube eliminating the need for transfer
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • Tubes have an 18-month shelf life from the date of manufacture
  • Nonscrew Safety Pull Caps can be removed with a gentle pull
Specifications
Chemical Identifiers
Lithium Heparin
Safety Pull Cap
Green with Yellow Ring
Specifications
Lithium Heparin
5 mL
13 x 100 mm
Safety Pull Cap
Green with Yellow Ring
Product Suggestions
Product Suggestions