Learn More
Greiner Bio-One Plasma Tubes with Lithium Heparin and Gel
For determinations in clinical chemistry and immunochemistry. Greiner Bio-One Plasma Tubes with Lithium Heparin and Gel includes a safety pull cap.
Supplier: Greiner Bio-One 455083P
Description
Plasma stability in a spun tube.
- Interior tube wall is coated with lithium heparin, an anticoagulant that binds antithrombins to block the coagulation cascade producing a plasma sample
- Contain a barrier gel in the tube that moves upward to form a stable barrier separating plasma from cells during centrifugation; plasma may be aspirated directly from the tube eliminating the need for transfer
- Clear as glass with equal or better performance
- Molded to be virtually unbreakable, providing a better level of safety than glass
- Tubes have an 18-month shelf life from the date of manufacture
- Nonscrew Safety Pull Caps can be removed with a gentle pull
Chemical Identifiers
| Lithium Heparin | |
| Safety Pull Cap | |
| Green with Yellow Ring |
Specifications
| Lithium Heparin | |
| 8 mL | |
| 16 x 100 mm |
| Safety Pull Cap | |
| Green with Yellow Ring |