missing translation for 'onlineSavingsMsg'
Learn More

Green Tea Extract, 4:1, Powder, Spectrum™ Chemical

Catalog No. p-7157500
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603056 25 g Amber Glass Bottle
18603057 100 g Amber Glass Bottle
18603058 500 g Amber Glass Bottle
18603059 1 kg Amber Glass Bottle
18603055 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18603056 Supplier Spectrum Chemical Mfg Cor Supplier No. G128125GM
Only null left

G1281, 84650-60-2

Spectrum™ Chemical Green Tea Extract, 4:1, Powder

Chemical Identifiers

CAS 84650-60-2

Specifications

CAS Max % 100%
Loss on Drying 9%
Packaging Amber Glass Bottle
Quantity 25 g
Grade Reagent