missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Green Tea Extract, 4:1, Powder, Spectrum™ Chemical
Description
Spectrum™ Chemical Green Tea Extract, 4:1, Powder
Specifications
Specifications
| CAS Max % | 100% |
| Loss on Drying | 9% |
| Packaging | Fiber Drum |
| Quantity | 25 kg |
| Grade | Reagent |