Learn More
Enzo Life Sciences BIOMOL Green (1x250ml) DFS Item
Supplier:  Enzo BMLAK1110250
A simple and convenient method for colorimetric phosphate quantitation. Applications: FUNC. Colorimetric detection, HTS. Contents: Biomol Green Reagent (Prod. No. BML-AK111) (1l or 250ml; liquid in screw cap plastic bottle) Storage: room temperature. Long-term at 4C; can be stored frozen without deleterious effects. Phosphate Standard (Prod. No. BML-KI102) (1ml; 800m phosphate in distilled water) Storage: room temperature. Long-term at 4C; can be stored frozen without deleterious effects.
Catalog No. NC9537789
Only null left
Explore available promotions