missing translation for 'onlineSavingsMsg'
Learn More

Graphite rod, 6.15mm (0.242in) dia x 305mm (12in) long, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog No. AA14739KM
Change view
Click to view available options
Quantity:
25 Pc.
Packaging:
Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA14739KM 25 Pc. Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA14739KM Supplier Thermo Scientific Chemicals Supplier No. 014739KM

 

Graphite rod is used in computer brushes, anodes, arc lamps, carbon coating, cathode ray tubes, and moderator in nuclear piles. It is used for high-temperature crucibles, as a lubricant and in lead pencils. Graphite rod coated with Th(IV), plays an useful application for ion selective electrode based on thorium oxinate complex. It is also essential for the preparation of lead(II)-selective PVC-coated graphite rod electrode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Graphite rod
Packaging Tube
Quantity 25 Pc.
MDL Number MFCD00144065
Solubility Information Insoluble in water. Soluble in molten nickel.
Percent Purity 99.9995%

RUO – Research Use Only