missing translation for 'onlineSavingsMsg'
Learn More

Graphite rod, 6.15mm (0.242 in.) dia x 305mm (12 in.) long, 99.9995% (metals basis), Thermo Scientific™

Catalog No. AA40768KM
Change view
Click to view available options
Quantity:
25 Pc.
Packaging:
Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA40768KM 25 Pc. Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA40768KM Supplier Thermo Scientific Chemicals Supplier No. 40768KM

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Name or Material Graphite rod
Packaging Tube
Quantity 25 Pc.
MDL Number MFCD00144065
Solubility Information Insoluble in water.
Percent Purity 99.9995%