missing translation for 'onlineSavingsMsg'
Learn More

Graphite rod, 5.0mm (0.20in) dia, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog No. AA45076DW
Change view
Click to view available options
Quantity:
300 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45076DW 300 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45076DW Supplier Thermo Scientific Chemicals Supplier No. 045076DW

 

Graphite rod is basically used for casting and polishing purposes, moulding & Carbon addition in steel industries. It is also used for smooth casting iron, copper base alloys and aluminum base alloy. It is mostly used for evaporating carbon onto specimens being prepared for either x-ray microanalysis (EDS or WDS) or transmission electron microscopy (TEM).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Graphite rod
Name Note 5.0mm (0.20 in.) dia.
Assay Percent Notes (metals basis)
Quantity 300 mm
MDL Number MFCD00144065
Solubility Information Insoluble in water.
Percent Purity 99.997%
Grade UF-4S

RUO – Research Use Only