missing translation for 'onlineSavingsMsg'
Learn More

Graphite plate, highly oriented pyrolytic graphite (HOPG), 10x10x1mm (0.394x0.394x0.039in), Thermo Scientific Chemicals

Catalog No. aa43834kj
Change view
Click to view available options
Quantity:
5 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43834KJ 5 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA43834KJ Supplier Thermo Scientific Chemicals Supplier No. 043834KJ
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Graphite plate
Packaging Shrink-wrapped
Quantity 5 Pc.
MDL Number MFCD00144065

RUO – Research Use Only