missing translation for 'onlineSavingsMsg'
Learn More

Graphite electrode, crater-drillpoint/undercut, 6.15mm dia, 38.10mm length, volume 0.072cc, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog No. AA14743KN
Change view
Click to view available options
Quantity:
100 Pc.
Packaging:
Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA14743KN 100 Pc. Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA14743KN Supplier Thermo Scientific Chemicals Supplier No. 014743KN
Only null left

 

Graphite electrodes are used mainly in electric arc furnace steel production. It also used to refine steel in ladle furnaces and in other smelting processes, furnance design, sharp requirements, water spray rings, charging practice.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Graphite electrode
Packaging Tube
Quantity 100 Pc.
Solubility Information Insoluble in water.
Percent Purity 99.9995%

RUO – Research Use Only