missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Graphic Controls Pen
Specifications
Specifications
| Product Type | Graphic Controls Pen |
| Color | Purple |
| For Use With (Application) | For chart recorders |
| Quantity | 6 Pk. |
| Length (English) | 0.6 in. |