missing translation for 'onlineSavingsMsg'
Learn More

Graphene Nanoplatelets 6-8nm(thick), 5micro m(wide), TCI America™

Catalog No. g04415g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G04415G Supplier TCI America Supplier No. G04415G
Only null left

Chemical Identifiers

CAS 1034343-98-0
Molecular Formula Cx
Molecular Weight (g/mol) 16.04
MDL Number MFCD00144065
InChI Key VNWKTOKETHGBQD-UHFFFAOYSA-N
Synonym methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon
PubChem CID 297
ChEBI CHEBI:16183
IUPAC Name Graphene
SMILES C

Specifications

Color Black
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Graphene Nanoplatelets 6-8nm(thick), 5micro m(wide)