missing translation for 'onlineSavingsMsg'
Learn More

Granisetron Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012403
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012403 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012403 Supplier Merck Emd Millipore Supplier No. PHR1616500MG
Only null left

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Pharmaceutical (small molecule)

Specifications

CAS 107007-99-8
Linear Formula C18H24N4 O · HCl
MDL Number MFCD01747034
Synonym Granisetron hydrochloride; 1-Methyl-N-[(3-endo)-9-methyl-9-azabicyclo[3.3.1 ]non-3-yl]-1 H-indazole-3-carboxamide hydrochloride
Type Certified Reference Material
Quantity 500 mg
Formula Weight 348.87
Grade Certified Reference Material