missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grams Iodine Solution, Certified, 0.025N ±0.001N, LabChem™
Supplier: LabChem LC149002
Specifications
| Liquid | |
| 7732-18-5,7681-11-0,7553-56-2,7647-01-0 | |
| Amber Glass | |
| Iodine vapor; Potassium oxide | |
| 1 L | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
| Certified |
| Grams Iodine Solution | |
| 99.02,0.64,0.32,0.02 | |
| Soluble in water | |
| Red to Brown | |
| I2 | |
| 253.809 | |
| 0.025N ±0.001N |
Chemical Identifiers
| 7732-18-5 | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |
Safety and Handling
GHS H Statement
Harmful to aquatic life.
GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage : Room Temperature