missing translation for 'onlineSavingsMsg'
Learn More

Grams Iodine Solution, Certified, 0.025N ±0.001N, LabChem™

Supplier:  LabChem LC149001

 View more versions of this product

Catalog No. LC149001


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Liquid
7732-18-5,7681-11-0,7553-56-2,7647-01-0
Amber Glass
Iodine vapor; Potassium oxide
500 mL
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Certified
Grams Iodine Solution
99.02,0.64,0.32,0.02
Soluble in water
Red to Brown
I2
253.809
0.025N ±0.001N

Chemical Identifiers

7732-18-5
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful to aquatic life.

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions