missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Gramicidin, 95%

Catalog No. p-5384788
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6677103 1 g
AAJ66771MC 100 mg
AAJ66771ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6677103 Supplier Thermo Scientific Chemicals Supplier No. J6677103
Only null left

For cellular and molecular biology applications | 1882.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Purification Method HPLC
Purity 95%
Content And Storage 2°C to 8°C
Format Powder
For Use With (Application) Targets primarily the cell membrane of gram positive bacteria and shows little effect against gram negative bacteria
Molecular Weight (g/mol) 1882.29
Product Type Gramicidin, 95%
Quantity 1 g
Regulatory Status RUO - research use only

RUO – Research Use Only