missing translation for 'onlineSavingsMsg'
Learn More

BD Gram Crystal Violet

Catalog No. B4312525
Change view
Click to view available options
Quantity:
3.8 L
4 x 250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4312525 4 x 250mL
B4312526 3.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4312525 Supplier BD Supplier No. 212525
Only null left

Can be used as a replacement reagent for the BD™ Gram Stain Kits. BD™ Gram Crystal Violet is used as the primary stain in a 4-step Gram staining process. Available in 250ml solution bottles for direct use or gallon containers for bulk storage.

Gram Stain reagents are used to stain microorganisms from cultures or specimens by the differential Gram method.

  • Complete stain kits reduce the mess associated with Gram staining procedures and include a compact tray for holding the 250mL reagent bottles
  • Break-resistant bottles features no-drip, keep-clean cap spouts that make regulating the flow easy and that help to prevent spots and spills
  • Color-coded labels allow quick identification of the reagent

Order Info

Available in packs of four 250mL bottles or individual one-gallon bottles

Specifications

Type Crystal Violet
Quantity 4 x 250mL
Certifications/Compliance CE
Format Liquid
For Use With (Application) Used to stain microorganisms from cultures or specimens by the differential gram method
Color Purple
Storage Requirements 15° to 30°C
Packaging Type Direct use reagent bottle
Stain Type Crystal Violet