Learn More
Eisco™ Graham Condensers
Catalog No. S23640
Change view
Click to view available options
Length (Metric) Jacket:
400 mm
300 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Jacket
S23640 300 mm
S27038 400 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S23640 Supplier Eisco™ Supplier No. CH0308HN
Only null left
Use to change substances from gas to liquid, made from heat resistant borosilicate glass.
  • Heat Resistant Borosilicate Glass
  • Type: Coiled Distillate
  • Available in two different lengths
Specifications
Standard Taper Joint No 29/32
Material Borosilicate Glass (3.3)
Length (Metric) Jacket 300 mm
For Use With (Application) Distillation
Type Coiled Distillate