Learn More
Synthware™ Graham Condensers
GREENER_CHOICE
Catalog No. 31500768
Change view
Click to view available options
Standard Taper Joint No:
14/20
19/22
24/40
Length (Metric) Jacket:
120 mm
200 mm
250 mm
300 mm
400 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No Length (Metric) Jacket
31500768 24/40 400 mm
31500767 24/40 300 mm
31500763 14/20 120 mm
31500764 19/22 200 mm
31500765 24/40 250 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 31500768 Supplier Synthware™ Supplier No. C144400
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Creates and captures vapor condensation created during a chemical reaction
  • With top standard taper outer joint and lower drip tip joint
Specifications
Material Borosilicate Glass (3.3)
Length (Metric) Jacket 400 mm
Standard Taper Joint No 24/40
For Use With (Application) Organic Synthesis
Type With hose connections
For Use With (Equipment) Round Bottom Flask