Learn More
Synthware™ Graham Condensers
Creates and captures vapor condensation created during a chemical reaction
Supplier: Synthware™ C149200
Description
- With top standard taper outer joint and lower drip tip joint
Chemical Identifiers
| Organic Synthesis |
Specifications
| Organic Synthesis | |
| Graham Condenser | |
| 200 mm |
| Glass | |
| 19/22 |