Learn More
United Scientific Supplies Graham Condensers, 24/40 Joint
Designed for use in distillation applications
Supplier:  United Scientific Supplies CNG500
 View more versions of this product
Catalog No. S43787
Only null left
Description
Description
  • Designed for use in distillation applications
  • Constructed of borosilicate glass
  • A coiled inner tube provides additional surface area for highly efficient cooling
  • Socket is 24/40 with a screw thread connector
Specifications
Chemical Identifiers
Screw Thread
Graham Condenser
Distillation Glassware
Borosilicate Glass
Distillation
Spiral
Specifications
Screw Thread
Graham Condenser
Distillation Glassware
500 mm
Borosilicate Glass
Distillation
Spiral
Product Suggestions
Product Suggestions