missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Graduated Cylinders, Cleaning Brushes, and Glass Droppers

Catalog No. S01893
Change view
Click to view available options
Product Type:
Labware Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S01893 Labware Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01893 Supplier Eisco Labs Supplier No. CH202002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes 4 borosilicate Class B cylinders (5 to 100 mL), 3 droppers, and 2 brushes. Durable, autoclavable, and chemical-resistant. Packed in foam for safe storage. A set of 9 Pieces.

  • A set of 9 Pieces: Set includes nine premium quality laboratory essentials - 4 graduated cylinders in 5 mL, 10 mL, 50 mL and 100 mL capacities, 3 droppers, and 2 cleaning brushes. Cylinders feature blue, printed graduations, round bases, and spouts.
  • Heavy Duty Glassware: Class B graduated cylinders made of high quality, heavy duty, borosilicate 3.3 glass. Thick walls provide strength against breakage.
  • Superior Durability: Autoclavable, with superior chemical durability and heat tolerance. Low temperature gradient as it can withstand high temperatures and thermal shocks. Designed for use in professional laboratories, but is an ideal choice for science classrooms and household use.
  • Safety Pack: Set is packaged in a custom-fit, thick foam casing, for safe storage and worry-free transport.
  • Set also includes two 1 mL ungraduated glass droppers, one 1 mL graduated dropper, and two cleaning brushes. Set is packaged in a custom-fit, thick foam casing, for secure storage and safe transport. Screen-printed packaging displays content and is shrink-wrapped to protect product from dust and fine debris.
  • Autoclavable at 121°C, with superior chemical durability.
  • Eisco Labs borosilicate glass has the following heat tolerances: Strain point - 515°C, Annealing point - 565°C and Softening point - 820°C.

Specifications

Grade 8 to 12
Material Borosilicate Glass
Class Size 1 Class
Product Type Labware Kit