missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Grade 25 Sapphire Spheres, Dia.: 3mm

$101.68

Specifications

Product Type Sapphire Sphere
Diameter (Metric) 3 mm
Grade Grade 25
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA42061KJ
View Documents Promotion Details
Thermo Scientific™
42061KJ
5 Pieces
Each for $101.68
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Sapphire Sphere
3 mm
Grade 25
Videos
SDS
Documents

Documents