missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Grade 25 Ruby Spheres, Dia.: 3.18mm

Catalog No. AA42044KM
Change view
Click to view available options
Quantity:
100 Pieces
25 Pieces
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42044KM 25 Pieces
AA42044KN 100 Pieces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42044KM Supplier Thermo Scientific™ Supplier No. 42044KM

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Product Type Mineral
Diameter (Metric) 3.18 mm
Grade Grade 25
Quantity 25 Pieces
Specimen Type Ruby Sphere