missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Grade 25 Ruby Spheres, Dia.: 1.5mm

Specifications

Product Type Ruby Sphere
Diameter (Metric) 1.5 mm
Grade Grade 25
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA42077KJ
View Documents Promotion Details
Thermo Scientific™
42077KJ
5 Pieces N/A
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ruby Sphere
1.5 mm
Grade 25
Videos
SDS
Documents

Documents