missing translation for 'onlineSavingsMsg'
Learn More

BUCHI FlashPure Flash Chromatography Cartridges

Catalog No. 05405565
Change view
Click to view available options
Bed Weight (Metric):
4 g
120 g
330 g
Includes:
10 Pc.
20 Pc.
4 Pc.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight (Metric) Includes
05405565 120 g 10 Pc.
05404811 4 g 20 Pc.
05405566 330 g 4 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05405565 Supplier BUCHI Supplier No. 145154067

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FlashPure flash chromatography cartridges offer high resolution and outstanding loading capacity.

  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster

Specifications

Material Silica - Normal Phase
Type FlashPure
Includes 10 Pc.
Resolution Medium, Irregular (35 to 45 μm)
Bed Weight (Metric) 120 g
For Use With (Application) Small Molecules
For Use With (Equipment) Pure Chromatography Systems