missing translation for 'onlineSavingsMsg'
Learn More

High-Tech Conversions Grab-EEZ™ Cleanroom Wipe Dispenser

Catalog No. 17444080
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17444080 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17444080 Supplier High-Tech Conversions Supplier No. GEDISP.99

Grab-EEZ™ Cleanroom Wipe Dispenser is an ISO Class 5 (Class 100) certified dispenser designed for use with the Grab-EEZ™ Cleanroom Wipes.

  • New and improved GRAB EEZ!
    • For use with 9 x 9 in. C-Folded Wipes.
    • Redesigned Cover Makes Loading Wipes Easier!
    • Optional Plastic Shield Protects Wipes Inside!
    • Still ESD safe, four holes for optional wall mounting, easy non cross contaminating dispensing, double bagged, and ready to bring into the cleanroom.
  • The GRAB-EEZ was designed with simplicity as a top priority:
    • Unique double C-folded cleanroom wipes, also manufactured by High-Tech Conversions are stacked inside the unit, protected from the environment until use. Each dispenser is prepared and cleaned in a cleanroom and double bagged before it is first introduced into the cleanroom where it will be used.
    • The GRAB-EEZ Cleanroom Wipe Dispenser is designed to reduce and eliminate cross contamination and wasted wipes by ensuring only one wipe is taken at a time. Typically, cleanroom wipes are supplied in flat sheets and placed on a table or in trays within a cleanroom but because the facility personnel is required to wear gloves, it is very difficult to select one wipe at a time.
  • Benefits of a cleanroom wipe dispenser include:
    • Easier To Use with Gloved Hands
    • Conforms To Good Housekeeping Practices
    • Counter Top or Wall Mountable
    • Unique Double C-Folded Design
    • Extremely Low in Particles and Low in Chemical Extractables
    • Eliminates wasteful wipe usage and cross-contamination

Specifications

Product Type Dispenser
Color White
Material Plastic
Certifications/Compliance ISO Cleanroom Class 5-6, USP 797 and 800 standards
For Use With (Equipment) Grab-EEZ™ Cleanroom Wipes, 9 x 9 in.