missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ GP Forward™

Catalog No. 17000042
Change view
Click to view available options
Volume (Metric):
3.78 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
17000042 3.78 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17000042 Supplier Diversey™ Supplier No. 101104494

A high performance cleaner for daily use on floors, walls and all washable surfaces

  • General purpose cleaner for counters, tables, floors, and other hard surfaces
  • Concentrated formulations deliver powerful cleaning performance for just pennies per diluted gallon
  • Removes all types of dirt, grit, grease, and soap scum with no rinsing required
  • Green in color with a pleasant citrus scent

Specifications

Product Type General Purpose Cleaner
pH 11.4
Sterility Non-sterile
Appearance Green
Certifications/Compliance Kosher, LONO
Container Type Jug
For Use With (Application) For general cleaning of any water-safe surface
Quantity 4 Pk.
Volume (English) 1 gal.
Volume (Metric) 3.78 L