Learn More
Promega GoTaq MDx Hot Start Polymerase, High Concentration, 1,000u
GoTaq MDx Hot Start Polymerase contains GoTaq MDx DNA Polymerase bound to a proprietary antibody that blocks polymerase activity.
Supplier:  Promega D6101
Encompass
Catalog No. PRD6101
Only null left
Description
Description
  • Inhibits Polymerase Activity at Temperatures below 70°C
  • Manufactured under cGMP
  • High−concentration and glycerol−free formulations also available
  • All formulations supplied at ≥5u/microl, except for high−concentration formulation supplied at ≥50u/microl
  • Custom formats and formulations
Specifications
Specifications
-30°C to -10°C
1000 U
Product Suggestions
Product Suggestions