Learn More
Promega GoTaq MDx Hot Start Polymerase, Glycerol-Free, 500u
Description
- Inhibits Polymerase Activity at Temperatures below 70°C
- Manufactured under cGMP
- High−concentration and glycerol−free formulations also available
- All formulations supplied at ≥5u/microl, except for high−concentration formulation supplied at ≥50u/microl
- Custom formats and formulations
Specifications
Specifications
| Quantity | 500 Units |