missing translation for 'onlineSavingsMsg'
Learn More

Promega GoTaq™ G2 Green Master Mix

GoTaq(R) G2 Master Mixes are premixed ready-to-use solutions containing GoTaq(R) G2 DNA Polymerase, dNTPs, MgCl2 and reaction buffers at optimal concentrations for efficient amplification of a wide range of DNA templates by PCR.

Supplier:  Promega M7822

Encompass

 View more versions of this product

Catalog No. PRM7822


Only null left

Description

Description

  • All-in-One PCR Master Mix: Just Add Template and Primers
  • High-Performance GoTaq(R) G2 DNA Polymerase in a Ready-to-Use Master Mix
  • Simplifies reaction setup and saves time
  • Use the green master mix for direct-to-gel analysis, and the colorless master mix when post-amplification fluorescence or absorbance analysis is required
Specifications

Chemical Identifiers

−30° to −10°C
Green Master Mix
RT-PCR

Specifications

−30° to −10°C
100 reactions
Green Master Mix (M782A 2 x 1.25mL), Nuclease-Free Water (P119A 2 x 1,250μL)
Green Master Mix
RT-PCR
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions