missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega GoTaq™ G2 Flexi DNA Polymerase
GoTaq™ G2 Flexi DNA Polymerase is supplied with 5X Green GoTaq™ Flexi Buffer and 5X Colorless GoTaq™ Flexi Buffer and 25mM MgCl2.
$34.04 - $1,688.72
Specifications
| Concentration | 5 U/μL |
|---|---|
| Content And Storage | −30° to −10°C |
| Description | Flexi DNA Polymerase |
| For Use With (Application) | RT-PCR |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PRM7805
|
Promega
M7805 |
500U |
Each for $116.39
|
|
|||||
PRM7801
|
Promega
M7801 |
100U |
Each for $34.04
|
|
|||||
PRM7808
|
Promega
M7808 |
10.000U |
Each for $1,688.72
|
|
|||||
PRM7806
|
Promega
M7806 |
2500U |
Each for $483.12
|
|
|||||
Description
- High-Performance GoTaq(R) G2 DNA Polymerase with Mg-Free Buffer System
- Full-length Taq DNA polymerase that exhibits 5′→3′ exonuclease activity
- Direct to gel convenience
- Flexibility to optimize Mg concentration in PCR
- Supplied at a concentration of 5u/μL
Specifications
| 5 U/μL | |
| −30° to −10°C | |
| Flexi DNA Polymerase | |
| RT-PCR |