missing translation for 'onlineSavingsMsg'
Learn More

Promega GoTaq™ Flexi DNA Polymerase

Taq DNA Polymerase. Provided with Green and Colorless Reaction Buffers without MgCl2, and 25mM MgCl2. Green Buffer allows direct gel analysis after amplification. Enhanced performance over standard Taq DNA Polymerase.

Supplier:  Promega M8296

Encompass

 View more versions of this product

Catalog No. PRM8296

Only null left

Includes: GoTaq Flexi DNA Polymerase, 5X Colorless GoTaq Flexi Buffer (Used when direct fluorescence or absorbance readings are required without prior purification of the DNA from the amplification reaction), 5X Green GoTaq Flexi Buffer (Contains two dyes that separate during electrophoresis to facilitate direct-to-gel analysis of PCR products), 25mM MgCl2

Description

Description

  • Optimize Enzyme and Magnesium Concentrations in Your PCR
  • Mg-free reaction buffers for ease of optimization
  • Direct-to-gel convenience
  • Supplied at a concentration of 5u/μL
Specifications

Chemical Identifiers

5 U/μL
DNA Polymerase
8
GoTaq Flexi DNA Polymerase, 5X Colorless GoTaq Flexi buffer (Used when direct fluorescence or absorbance readings are required without prior purification of the DNA from the amplification reaction), 5X Green GoTaq Flexi buffer, 25mM MgCl2
−30 to −10°C
Liquid
Standard PCR

Specifications

5 U/μL
DNA Polymerase
8
Standard PCR
−30 to −10°C
Liquid
2500U
GoTaq Flexi DNA Polymerase, 5X Colorless GoTaq Flexi buffer (Used when direct fluorescence or absorbance readings are required without prior purification of the DNA from the amplification reaction), 5X Green GoTaq Flexi buffer, 25mM MgCl2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions