missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega GoTaq™ Flexi DNA Polymerase
Description
Includes
GoTaq Flexi DNA Polymerase, 5X Colorless GoTaq Flexi Buffer (Used when direct fluorescence or absorbance readings are required without prior purification of the DNA from the amplification reaction), 5X Green GoTaq Flexi Buffer (Contains two dyes that separate during electrophoresis to facilitate direct-to-gel analysis of PCR products), 25mM MgCl2
- Optimize Enzyme and Magnesium Concentrations in Your PCR
- Mg-free reaction buffers for ease of optimization
- Direct-to-gel convenience
- Supplied at a concentration of 5u/μL
- The purchase of this product conveys to the buyer the limited, nonexclusive, nontransferable right (without the right to resell, repackage, or further sublicense) under U.S. Published Patent Appln. 20020150900 and U.S. Pat. Nos. 5,432,272, 6,617,106 and 6,140,496 to use the product. No other license is granted to the buyer whether expressly, by implication, by estoppel or otherwise. In particular, the purchase of this product does not include or carry any right or license to sell this product. For information on purchasing a license for other uses, please contact Promega Corporation, Licensing, 2800 Woods Hollow Road, Madison, WI 53711, or EraGen Biosciences, Corporate Licensing, 918 Deming Way, Suite 201, Madison, WI 53717. Phone (608) 662-9000; Fax (608) 662-9003.
- This product is designed and sold for use in the multiplex PCR process covered by U.S. Pat. No. 5,582,989 and Canadian Pat. No. 1,339,731. A limited license has been granted under the patent to use only this amount of the product to practice the multiplex PCR process and is conveyed to the purchaser by the purchase of this product.
- Patents for the foundational PCR process, European Pat. Nos. 201,184 and 200,362, expired on March 28, 2006. In the U.S., the patents covering the foundational PCR process expired on March 29, 2005.
Specifications
Specifications
| Concentration | 5 U/μL |
| Content And Storage | −30 to −10°C |
| Description | DNA Polymerase |
| Form | Liquid |
| pH | 8 |
| Quantity | 10000U |
| For Use With (Application) | Standard PCR |
| Includes | GoTaq Flexi DNA Polymerase, 5X Colorless GoTaq Flexi buffer (Used when direct fluorescence or absorbance readings are required without prior purification of the DNA from the amplification reaction), 5X Green GoTaq Flexi buffer, 25mM MgCl2 |