missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Autoclave Bags Printed with Biohazard Logo

Catalog No. 07202044
Change view
Click to view available options
Capacity (Metric):
3 L
9 L
14 L
21 L
33 L
Width (Metric):
255 mm
300 mm
400 mm
600 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Width (Metric)
07202044 9 L 300 mm
07202043 3 L 255 mm
07202045 14 L 400 mm
07202046 21 L 400 mm
07202047 33 L 600 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 07202044 Supplier Corning™ Supplier No. SA0910
Only null left

Used for contaminated waste disposal in autoclave. Gosselin™ Autoclave Bags Printed with Biohazard Logo can withstand autoclaving temperatures up to 134°C/273.2°F.

  • Polypropylene (PP) resistant bags
  • Thermal resistance 134°C
  • Transparent

Specifications

Material Polypropylene
No. per Case 500/Cs.
No. per Pack 1/Pk.
Height (Metric) 500 mm
Capacity (Metric) 9 L
Width (Metric) 300 mm
Thickness (Metric) 40 μm