missing translation for 'onlineSavingsMsg'
Learn More
Learn More
St Martin Press Golden Guides
Description
- Golden Guides have been a science classroom staple for years
- Full of colorful illustrations and informative easy-to-read text
- Paperback, 4 x 6 in.
Specifications
Specifications
| Format | Paperback Book |
| Product Type | Whales, Dolphins and Other Marine Mammals |
| For Use With (Application) | Full of colorful, magnificently illustrated guide gives information on natural history and evolution of these fascinating creatures, distribution and migratory patterns |
| Subject | Biology |
| Author | George S. Fichter |