Learn More
New England Biolab Ltd NEB Golden Gate Enzyme Kit (BsaI-HFv2) 100 reactions
Catalog No. NC1576745
Encompass
The NEB Golden Gate Assembly Kit (BsaI-HFv2) contains an optimized mix of BsaI-HFv2 and T4 DNA Ligase. Together these enzymes can direct the assembly of multiple inserts/modules using the Golden Gate approach. Also included is the pGGAselectdestination plasmid, which provides a backbone for your assembly, features convenient restriction enzyme sites for subcloning, and has T7/SP6 promoter sequences to enable in vitro transcription. Please note that while general descriptions regarding Golden Gate Assembly use the BsaI nomenclature, this kit and protocols feature the specific engineered form optimized for Golden Gate Assembly, BsaI-HFv2.
Catalog No. NC1576745 Supplier New England Biolab Ltd Supplier No. E1601L
The NEB Golden Gate Assembly Kit (BsaI-HFv2) contains an optimized mix of BsaI-HFv2 and T4 DNA Ligase. Together these enzymes can direct the assembly of multiple inserts/modules using the Golden Gate approach. Also included is the pGGAselectdestination plasmid, which provides a backbone for your assembly, features convenient restriction enzyme sites for subcloning, and has T7/SP6 promoter sequences to enable in vitro transcription. Please note that while general descriptions regarding Golden Gate Assembly use the BsaI nomenclature, this kit and protocols feature the specific engineered form optimized for Golden Gate Assembly, BsaI-HFv2.