missing translation for 'onlineSavingsMsg'
Learn More

Gold's Reagent 97.0+%, TCI America™

Catalog No. p-7130255
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03935G 5 g
G039325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G03935G Supplier TCI America Supplier No. G03935G
Only null left

Chemical Identifiers

CAS 20353-93-9
Molecular Formula C6H14ClN3
Molecular Weight (g/mol) 163.649
MDL Number MFCD00011793
InChI Key DEIBXAPEZDJDRC-UHFFFAOYSA-M
Synonym [[[(Dimethylamino)methylene]amino]methylene]dimethylammonium Chloride
PubChem CID 11008255
IUPAC Name (dimethylaminomethylideneamino)methylidene-dimethylazanium;chloride
SMILES CN(C)C=NC=[N+](C)C.[Cl-]

Specifications

Melting Point 99°C
Color Yellow
Quantity 5 g
Formula Weight 163.65
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Gold′s Reagent