Learn More
MilliporeSigma™ Supelco™ Gold-Plated Inlet Seals
Description
- Replacement inlet seals are manufactured using high quality stainless steel plated with pure gold to insure inertness
- No brighteners are used in the plating process
- Each pack includes one washer for each seal
Specifications
Specifications
| For Use With (Application) | Suitable for Applications with Low Split Flows (< 200 mL/min.) |
| Includes | One Washer for Each Seal |
| Material | Stainless Steel |
| Quantity | 2/Pk. |
| Description | Gold-Plated |
| Type | Straight Design |