missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Gold Leaves Pack

Catalog No. S24896
Change view
Click to view available options
Class Size:
25 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S24896 25 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24896 Supplier Eisco™ Supplier No. PH0914

Gold Leaves Pack for use with electroscope.

  • For spare use
  • In plastic case

Specifications

Color Gold
Grade 9 to 12
Dimensions (L x W) 1.25 x 0.5 in.
For Use With (Application) Studying basic electrostatic principles when used in an electroscope
For Use With (Equipment) Electroscope
Includes 6 Leaves, 1 Plastic Case
Material Gold
Quantity 6
Width (English) 0.5 in.
Width (Metric) 12.7 mm
Class Size 25 Students
Product Type Gold Leaves
Description Gold Leaves
Show More Show Less